siRNA Details
Gene Symbol: R3HDML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178491.3 | NP_848586.1 | 2 | 330 | ||
GenBank mRNA | BC107048.2 | - | 2 | 330 | 40003509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
