siRNA Details
Gene Symbol: FZR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136197.1 | NP_001129669.1 | 2, 3 | 228 | ||
NM_001136198.1 | NP_001129670.1 | 2, 3 | 228 | |||
NM_016263.3 | NP_057347.2 | 3, 4 | 406 | |||
XM_005259573.4 | XP_005259630.1 | 3, 4 | 588 | |||
XM_017026863.1 | XP_016882352.1 | 4, 5 | 932 | |||
GenBank mRNA | AB013462.1 | - | 2, 3 | 194 | ||
AB013463.1 | - | 2, 3 | 194 | |||
AB033068.1 | - | 2, 3 | 228 | |||
AF080397.1 | - | 2, 3 | 206 | |||
AF083810.1 | - | 3, 4 | 333 | |||
AF102507.1 | - | 3, 4 | 335 | |||
AF433157.1 | - | 2, 3 | 194 | |||
BC013413.2 | - | 3, 4 | 383 | 3160334 | ||
BT007115.1 | - | 2, 3 | 194 | |||
DB462260.1 | - | 3, 4 | 406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
