siRNA Details
Gene Symbol: FZR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136197.1 | NP_001129669.1 | 11 | 2026 | ||
NM_001136198.1 | NP_001129670.1 | 13 | 2302 | |||
NM_016263.3 | NP_057347.2 | 14 | 2471 | |||
XM_005259573.4 | XP_005259630.1 | 14 | 2662 | |||
XM_017026863.1 | XP_016882352.1 | 15 | 3006 | |||
GenBank mRNA | AB033068.1 | - | 13 | 2294 | ||
AF083810.1 | - | 14 | 2398 | |||
AF102508.1 | - | 4 | 1208 | |||
BC111760.1 | - | 1 | 774 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
