siRNA Details
Gene Symbol: FZR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136197.1 | NP_001129669.1 | 11 | 1235 | ||
NM_001136198.1 | NP_001129670.1 | 13 | 1511 | |||
NM_016263.3 | NP_057347.2 | 14 | 1680 | |||
XM_005259573.4 | XP_005259630.1 | 14 | 1871 | |||
XM_017026863.1 | XP_016882352.1 | 15 | 2215 | |||
GenBank mRNA | AB013462.1 | - | 13 | 1477 | ||
AB013463.1 | - | 11 | 1201 | |||
AB033068.1 | - | 13 | 1502 | |||
AF080397.1 | - | 13 | 1480 | |||
AF083810.1 | - | 14 | 1607 | |||
AF102508.1 | - | 4 | 417 | |||
AF433157.1 | - | 11 | 1201 | |||
BC013413.2 | - | 14 | 1657 | 3160334 | ||
BT007115.1 | - | 13 | 1468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
