siRNA Details
Gene Symbol: COA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321197.1 | NP_001308126.1 | 7 | 1019 | ||
NM_001321198.1 | NP_001308127.1 | 7 | 900 | |||
NM_001321199.1 | NP_001308128.1 | 7 | 864 | |||
NM_001321200.1 | NP_001308129.1 | 7 | 920 | |||
NM_001321201.1 | NP_001308130.1 | 6 | 792 | |||
NM_018224.3 | NP_060694.2 | 6 | 799 | |||
XM_017012423.1 | XP_016867912.1 | 7 | 823 | |||
XM_017012425.1 | XP_016867914.1 | 7 | 907 | |||
XM_017012426.1 | XP_016867915.1 | 7 | 1026 | |||
XM_017012427.1 | XP_016867916.1 | 8 | 981 | |||
XM_017012428.1 | XP_016867917.1 | 6 | 806 | |||
XM_017012429.1 | XP_016867918.1 | 5 | 700 | |||
GenBank mRNA | AK021905.1 | - | 7 | 969 | ||
AK097203.1 | - | 5 | 2330 | |||
AK129690.1 | - | 3 | 1523 | |||
BC056884.1 | - | 7 | 791 | 6159045 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
