siRNA Details
Gene Symbol: SPATA6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716795.2 | XP_006716858.1 | 2, 3 | 281 | ||
XM_006716798.3 | XP_006716861.1 | 3, 4 | 286 | |||
XM_006716799.3 | XP_006716862.1 | 2, 3 | 388 | |||
XM_006716800.2 | XP_006716863.1 | 2, 3 | 236 | |||
XM_011517952.1 | XP_011516254.1 | 2, 3 | 224 | |||
XM_011517956.1 | XP_011516258.1 | 2, 3 | 281 | |||
XM_011517958.1 | XP_011516260.1 | 1, 2 | 142 | |||
XM_017014881.1 | XP_016870370.1 | 2, 3 | 281 | |||
XM_017014882.1 | XP_016870371.1 | 2, 3 | 281 | |||
XM_017014883.1 | XP_016870372.1 | 2, 3 | 281 | |||
XM_017014884.1 | XP_016870373.1 | 2, 3 | 236 | |||
XM_017014885.1 | XP_016870374.1 | 2, 3 | 281 | |||
GenBank mRNA | AK000920.1 | - | 3, 4 | 262 | ||
AK301790.1 | - | 2, 3 | 381 | |||
BC034293.1 | - | 2, 3 | 355 | 5167647 | ||
BC036349.1 | - | 3, 4 | 280 | |||
BX649030.1 | - | 2, 3 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
