siRNA Details
Gene Symbol: SPATA6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039395.3 | NP_001034484.3 | 3 | 306 | ||
XM_006716795.2 | XP_006716858.1 | 2 | 159 | |||
XM_006716798.3 | XP_006716861.1 | 3 | 164 | |||
XM_006716799.3 | XP_006716862.1 | 2 | 266 | |||
XM_006716800.2 | XP_006716863.1 | 2 | 114 | |||
XM_006716801.2 | XP_006716864.1 | 3 | 306 | |||
XM_011517952.1 | XP_011516254.1 | 2 | 102 | |||
XM_011517954.1 | XP_011516256.1 | 2 | 246 | |||
XM_011517956.1 | XP_011516258.1 | 2 | 159 | |||
XM_011517957.1 | XP_011516259.1 | 2 | 246 | |||
XM_011517958.1 | XP_011516260.1 | 1 | 20 | |||
XM_017014881.1 | XP_016870370.1 | 2 | 159 | |||
XM_017014882.1 | XP_016870371.1 | 2 | 159 | |||
XM_017014883.1 | XP_016870372.1 | 2 | 159 | |||
XM_017014884.1 | XP_016870373.1 | 2 | 114 | |||
XM_017014885.1 | XP_016870374.1 | 2 | 159 | |||
XM_017014886.1 | XP_016870375.1 | 2 | 62 | |||
XM_017014887.1 | XP_016870376.1 | 2 | 144 | |||
GenBank mRNA | AK000920.1 | - | 3 | 140 | ||
AK295831.1 | - | 3 | 298 | |||
AK301790.1 | - | 2 | 259 | |||
BC034293.1 | - | 2 | 233 | 5167647 | ||
BC036349.1 | - | 3 | 158 | |||
BX649030.1 | - | 2 | 209 | |||
DA234263.1 | - | 3 | 306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
