siRNA Details
Gene Symbol: SYNGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320523.1 | NP_001307452.1 | 3 | 1259 | ||
NM_004710.4 | NP_004701.1 | 4 | 1399 | |||
XM_005257792.3 | XP_005257849.1 | 3 | 1482 | |||
GenBank mRNA | AJ002308.1 | - | 4 | 1356 | ||
AL833010.1 | - | 1 | 2905 | |||
AY358916.1 | - | 4 | 1377 | |||
BC000407.2 | - | 4 | 1336 | 2823026 | ||
BC029755.1 | - | 4 | 1332 | 4341106 | ||
BC105992.1 | - | 3 | 1422 | 4746277 | ||
BI160076.1 | - | 1 | 569 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
