siRNA Details
Gene Symbol: SYNGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320523.1 | NP_001307452.1 | 3 | 744 | ||
NM_004710.4 | NP_004701.1 | 4 | 884 | |||
XM_005257792.3 | XP_005257849.1 | 3 | 967 | |||
GenBank mRNA | AJ002308.1 | - | 4 | 841 | ||
AL833010.1 | - | 1 | 2390 | |||
AY358916.1 | - | 4 | 862 | |||
BC000407.2 | - | 4 | 821 | 2823026 | ||
BC029755.1 | - | 4 | 817 | 4341106 | ||
BC105992.1 | - | 3 | 907 | 4746277 | ||
BI160076.1 | - | 1 | 54 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
