siRNA Details
Gene Symbol: MCM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270472.1 | NP_001257401.1 | 11 | 1878 | ||
NM_002388.4 | NP_002379.3 | 12 | 1991 | |||
GenBank mRNA | AK222586.1 | - | 12 | 1829 | ||
AK299566.1 | - | 11 | 1717 | |||
AK300754.1 | - | 11 | 1704 | |||
AK301704.1 | - | 12 | 1930 | |||
AK312928.1 | - | 12 | 1800 | |||
AY032603.1 | - | 12 | 1837 | |||
BC001626.1 | - | 12 | 1836 | 3542098 | ||
BC003509.1 | - | 12 | 1837 | 3610786 | ||
D38073.1 | - | 12 | 1861 | |||
KU178170.1 | - | 12 | 1784 | |||
KU178171.1 | - | 12 | 1784 | |||
X62153.2 | - | 12 | 1828 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
