siRNA Details
Gene Symbol: SOBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018013.3 | NP_060483.3 | 6 | 1727 | ||
XM_005267041.4 | XP_005267098.1 | 7 | 1888 | |||
XM_005267042.4 | XP_005267099.1 | 6 | 1793 | |||
XM_011535920.2 | XP_011534222.1 | 7 | 1888 | |||
XM_011535921.2 | XP_011534223.1 | 7 | 1412 | |||
XM_011535923.2 | XP_011534225.1 | 2 | 626 | |||
XM_017010991.1 | XP_016866480.1 | 5 | 2250 | |||
XM_017010992.1 | XP_016866481.1 | 5 | 1019 | |||
XM_017010993.1 | XP_016866482.1 | 5 | 1049 | |||
XM_017010994.1 | XP_016866483.1 | 5 | 1008 | |||
GenBank mRNA | BC113966.1 | - | 1 | 13 | ||
DQ507800.1 | - | 6 | 1223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
