siRNA Details
Gene Symbol: TTC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022492.4 | NP_071937.4 | 13 | 1492 | ||
NR_027749.1 | 12 | 1383 | ||||
XM_005264492.1 | XP_005264549.1 | 13 | 1475 | |||
XM_011533035.1 | XP_011531337.1 | 13 | 1541 | |||
XM_011533036.1 | XP_011531338.1 | 13 | 1538 | |||
XM_011533037.1 | XP_011531339.1 | 13 | 1535 | |||
XM_011533040.1 | XP_011531342.1 | 12 | 1490 | |||
XM_017004706.1 | XP_016860195.1 | 13 | 1478 | |||
XM_017004707.1 | XP_016860196.1 | 13 | 1472 | |||
XM_017004708.1 | XP_016860197.1 | 12 | 1386 | |||
XM_017004709.1 | XP_016860198.1 | 12 | 1610 | |||
GenBank mRNA | AK022850.1 | - | 9 | 1002 | ||
AK026819.1 | - | 1 | 478 | |||
AK301585.1 | - | 6 | 547 | |||
AK303105.1 | - | 8 | 783 | |||
AK303245.1 | - | 5 | 474 | |||
BC036017.1 | - | 13 | 1476 | 5270575 | ||
BC045536.1 | - | 12 | 1374 | |||
BX648131.1 | - | 7 | 2428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
