siRNA Details
Gene Symbol: TTC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022492.4 | NP_071937.4 | 8 | 843 | ||
NR_027749.1 | 7 | 740 | ||||
XM_005264492.1 | XP_005264549.1 | 8 | 832 | |||
XM_011533035.1 | XP_011531337.1 | 8 | 832 | |||
XM_011533036.1 | XP_011531338.1 | 8 | 832 | |||
XM_011533037.1 | XP_011531339.1 | 8 | 832 | |||
XM_011533040.1 | XP_011531342.1 | 7 | 781 | |||
XM_017004706.1 | XP_016860195.1 | 8 | 832 | |||
XM_017004707.1 | XP_016860196.1 | 8 | 832 | |||
XM_017004708.1 | XP_016860197.1 | 7 | 737 | |||
XM_017004709.1 | XP_016860198.1 | 7 | 967 | |||
GenBank mRNA | AK022850.1 | - | 8 | 829 | ||
AK090520.1 | - | 7 | 882 | |||
BC036017.1 | - | 8 | 827 | 5270575 | ||
BC045536.1 | - | 7 | 731 | |||
BC096709.1 | - | 8 | 829 | 40023059 | ||
BC098365.1 | - | 8 | 829 | 40023060 | ||
BC099729.1 | - | 6 | 1114 | |||
BX648131.1 | - | 5 | 1295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
