siRNA Details
Gene Symbol: DDX55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020936.2 | NP_065987.1 | 14 | 1871 | ||
NR_135104.1 | 14 | 1943 | ||||
NR_135105.1 | 14 | 1855 | ||||
XM_017019710.1 | XP_016875199.1 | 14 | 1910 | |||
XM_017019711.1 | XP_016875200.1 | 13 | 1965 | |||
XM_017019712.1 | XP_016875201.1 | 14 | 1763 | |||
XM_017019713.1 | XP_016875202.1 | 14 | 1886 | |||
XM_017019714.1 | XP_016875203.1 | 15 | 2044 | |||
XM_017019715.1 | XP_016875204.1 | 12 | 1611 | |||
XM_017019716.1 | XP_016875205.1 | 12 | 1556 | |||
XM_017019717.1 | XP_016875206.1 | 14 | 1808 | |||
XM_017019718.1 | XP_016875207.1 | 12 | 2226 | |||
XM_017019719.1 | XP_016875208.1 | 14 | 1763 | |||
XM_017019720.1 | XP_016875209.1 | 12 | 2841 | |||
GenBank mRNA | AB046815.1 | - | 9 | 3698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
