siRNA Details
Gene Symbol: DDX55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020936.2 | NP_065987.1 | 13 | 1429 | ||
NR_135104.1 | 13 | 1501 | ||||
XM_011538608.2 | XP_011536910.1 | 11 | 1187 | |||
XM_017019710.1 | XP_016875199.1 | 13 | 1468 | |||
XM_017019711.1 | XP_016875200.1 | 12 | 1523 | |||
XM_017019712.1 | XP_016875201.1 | 13 | 1321 | |||
XM_017019713.1 | XP_016875202.1 | 13 | 1444 | |||
XM_017019714.1 | XP_016875203.1 | 14 | 1602 | |||
XM_017019717.1 | XP_016875206.1 | 13 | 1366 | |||
XM_017019718.1 | XP_016875207.1 | 11 | 1784 | |||
XM_017019719.1 | XP_016875208.1 | 13 | 1321 | |||
XM_017019720.1 | XP_016875209.1 | 11 | 2399 | |||
XM_017019721.1 | XP_016875210.1 | 11 | 1131 | |||
GenBank mRNA | AB046815.1 | - | 8 | 3256 | ||
AK307384.1 | - | 13 | 1405 | |||
BC030020.2 | - | 13 | 1373 | 4821394 | ||
BC035911.1 | - | 2 | 278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
