siRNA Details
Gene Symbol: DDX55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020936.2 | NP_065987.1 | 6 | 532 | ||
NR_135104.1 | 6 | 532 | ||||
NR_135105.1 | 6 | 532 | ||||
XM_011538608.2 | XP_011536910.1 | 6 | 520 | |||
XM_017019710.1 | XP_016875199.1 | 6 | 517 | |||
XM_017019711.1 | XP_016875200.1 | 5 | 572 | |||
XM_017019712.1 | XP_016875201.1 | 6 | 370 | |||
XM_017019713.1 | XP_016875202.1 | 6 | 547 | |||
XM_017019714.1 | XP_016875203.1 | 7 | 705 | |||
XM_017019715.1 | XP_016875204.1 | 6 | 518 | |||
XM_017019716.1 | XP_016875205.1 | 6 | 517 | |||
XM_017019717.1 | XP_016875206.1 | 6 | 479 | |||
XM_017019718.1 | XP_016875207.1 | 4 | 897 | |||
XM_017019719.1 | XP_016875208.1 | 6 | 488 | |||
XM_017019720.1 | XP_016875209.1 | 4 | 1566 | |||
XM_017019721.1 | XP_016875210.1 | 6 | 518 | |||
GenBank mRNA | AB046815.1 | - | 1 | 2359 | ||
AK074219.1 | - | 6 | 484 | |||
AK131442.1 | - | 3 | 771 | |||
AK301043.1 | - | 6 | 495 | |||
AK307384.1 | - | 6 | 508 | |||
AL833756.1 | - | 4 | 1569 | |||
BC030020.2 | - | 6 | 476 | 4821394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
