siRNA Details
Gene Symbol: IRF2BPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024496.3 | NP_078772.1 | 1 | 1151 | ||
GenBank mRNA | BC108292.1 | - | 1 | 1151 | ||
BM744266.1 | - | 1 | 319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
