siRNA Details
Gene Symbol: SLC25A52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034172.2 | NP_001029344.3 | 1 | 740 | ||
GenBank mRNA | AK302337.1 | - | 1 | 740 | ||
BC103999.1 | - | 1 | 596 | 40023323 | ||
BC104000.1 | - | 1 | 596 | 40023324 | ||
BC104001.1 | - | 1 | 596 | 40023325 |
Gene Symbol: SLC25A51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033412.3 | NP_219480.1 | 3 | 971 | ||
GenBank mRNA | BC008500.1 | - | 3 | 732 | 4286914 |
Gene Symbol: SLC25A51P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026540.1 | 1 | 1056 | |||
GenBank mRNA | BC024198.1 | - | 1 | 1057 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
