siRNA Details
Gene Symbol: ZC3H13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015070.4 | NP_055885.3 | 14 | 3246 | ||
XM_005266301.3 | XP_005266358.1 | 15 | 3624 | |||
XM_005266302.3 | XP_005266359.1 | 15 | 3324 | |||
XM_005266303.3 | XP_005266360.1 | 16 | 3649 | |||
XM_005266304.3 | XP_005266361.1 | 15 | 3606 | |||
XM_005266305.3 | XP_005266362.1 | 16 | 3642 | |||
XM_005266306.3 | XP_005266363.1 | 15 | 3320 | |||
XM_005266307.3 | XP_005266364.1 | 15 | 3624 | |||
XM_005266308.3 | XP_005266365.1 | 14 | 3540 | |||
XM_005266309.3 | XP_005266366.1 | 14 | 3240 | |||
XM_005266310.3 | XP_005266367.1 | 14 | 3522 | |||
XM_005266311.3 | XP_005266368.1 | 14 | 3540 | |||
XM_005266312.1 | XP_005266369.1 | 14 | 3540 | |||
XM_017020475.1 | XP_016875964.1 | 16 | 3412 | |||
XM_017020476.1 | XP_016875965.1 | 16 | 3409 | |||
XM_017020477.1 | XP_016875966.1 | 15 | 3565 | |||
XM_017020478.1 | XP_016875967.1 | 15 | 3558 | |||
XM_017020479.1 | XP_016875968.1 | 15 | 3328 | |||
XM_017020480.1 | XP_016875969.1 | 14 | 3236 | |||
XM_017020481.1 | XP_016875970.1 | 15 | 3565 | |||
XM_017020482.1 | XP_016875971.1 | 14 | 3240 | |||
GenBank mRNA | AB020660.1 | - | 3 | 1072 | ||
AK074924.1 | - | 1 | 300 | |||
AL136745.1 | - | 7 | 2282 | |||
AY283618.1 | - | 14 | 3244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
