siRNA Details
Gene Symbol: CHD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032221.4 | NP_115597.3 | 13 | 1907 | ||
XM_005260573.2 | XP_005260630.1 | 12 | 1729 | |||
XM_011529080.2 | XP_011527382.1 | 12 | 1968 | |||
XM_011529082.2 | XP_011527384.1 | 1 | 36 | |||
XM_017028099.1 | XP_016883588.1 | 12 | 3403 | |||
XM_017028100.1 | XP_016883589.1 | 12 | 1928 | |||
XM_017028101.1 | XP_016883590.1 | 12 | 3403 | |||
XM_017028102.1 | XP_016883591.1 | 12 | 3402 | |||
XM_017028103.1 | XP_016883592.1 | 10 | 1362 | |||
XM_017028104.1 | XP_016883593.1 | 12 | 3403 | |||
GenBank mRNA | AK296955.1 | - | 12 | 1737 | ||
AY034072.1 | - | 13 | 1907 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
