siRNA Details
Gene Symbol: CHD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032221.4 | NP_115597.3 | 11 | 1551 | ||
XM_005260573.2 | XP_005260630.1 | 10 | 1373 | |||
XM_011529080.2 | XP_011527382.1 | 10 | 1612 | |||
XM_017028099.1 | XP_016883588.1 | 10 | 3047 | |||
XM_017028100.1 | XP_016883589.1 | 10 | 1572 | |||
XM_017028101.1 | XP_016883590.1 | 10 | 3047 | |||
XM_017028102.1 | XP_016883591.1 | 10 | 3046 | |||
XM_017028103.1 | XP_016883592.1 | 8 | 1006 | |||
XM_017028104.1 | XP_016883593.1 | 10 | 3047 | |||
GenBank mRNA | AF525085.1 | - | 6 | 481 | ||
AK296955.1 | - | 10 | 1381 | |||
AY034072.1 | - | 11 | 1551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
