siRNA Details
Gene Symbol: SRSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003017.4 | NP_003008.1 | 2 | 309 | ||
NR_036610.1 | 2 | 309 | ||||
GenBank mRNA | AB451229.1 | - | 1 | 138 | ||
AB451352.1 | - | 1 | 138 | |||
AF107405.1 | - | 2 | 233 | |||
AK304097.1 | - | 2 | 256 | |||
AK310317.1 | - | 887 | ||||
AK312553.1 | - | 2 | 260 | |||
BC000914.1 | - | 2 | 243 | 3049167 | ||
BC069018.1 | - | 2 | 239 | 3507371 | ||
BT007017.1 | - | 1 | 138 | |||
DC398250.1 | - | 2 | 401 | |||
L10838.1 | - | 1 | 138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
