siRNA Details
Gene Symbol: NXPE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077639.1 | NP_001071107.1 | 5 | 1181 | ||
NM_017678.2 | NP_060148.2 | 5 | 717 | |||
XM_011542881.2 | XP_011541183.1 | 5 | 1488 | |||
GenBank mRNA | AK000134.1 | - | 5 | 705 | ||
AK225236.1 | - | 5 | 705 | |||
AY358805.1 | - | 5 | 1097 | |||
AY547307.1 | - | 3 | 286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
