siRNA Details
Gene Symbol: EMILIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032048.2 | NP_114437.2 | 8 | 3688 | ||
GenBank mRNA | AB026706.1 | - | 4 | 994 | ||
AF270513.1 | - | 8 | 3688 | |||
AK090519.1 | - | 5 | 1783 | |||
BC032231.1 | - | 6 | 1361 | |||
BC136541.1 | - | 8 | 3642 | 9020531 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
