siRNA Details
Gene Symbol: ANKRD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286771.2 | NP_001273700.1 | 5 | 720 | ||
NM_015574.1 | NP_056389.1 | 5 | 1102 | |||
NM_032217.4 | NP_115593.3 | 5 | 1102 | |||
NM_198889.2 | NP_942592.1 | 5 | 1102 | |||
XM_005265671.3 | XP_005265728.1 | 5 | 1446 | |||
XM_005265672.3 | XP_005265729.1 | 5 | 1446 | |||
XM_005265673.3 | XP_005265730.1 | 5 | 1446 | |||
XM_017008011.1 | XP_016863500.1 | 5 | 677 | |||
XM_017008012.1 | XP_016863501.1 | 5 | 677 | |||
XM_017008013.1 | XP_016863502.1 | 5 | 677 | |||
XM_017008014.1 | XP_016863503.1 | 5 | 1446 | |||
XM_017008015.1 | XP_016863504.1 | 5 | 677 | |||
XM_017008016.1 | XP_016863505.1 | 5 | 1446 | |||
XM_017008017.1 | XP_016863506.1 | 5 | 677 | |||
GenBank mRNA | AB014597.2 | - | 5 | 626 | ||
AF308285.1 | - | 5 | 1089 | |||
AK300302.1 | - | 5 | 676 | |||
AK307581.1 | - | 5 | 1102 | |||
AL833287.1 | - | 2 | 279 | |||
BC004173.2 | - | 5 | 1083 | |||
BC007747.2 | - | 3 | 432 | |||
BC043394.1 | - | 5 | 1071 | |||
BX648035.1 | - | 5 | 675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
