siRNA Details
Gene Symbol: ANKRD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286771.2 | NP_001273700.1 | 8 | 1151 | ||
NM_015574.1 | NP_056389.1 | 8 | 1533 | |||
NM_032217.4 | NP_115593.3 | 8 | 1533 | |||
NM_198889.2 | NP_942592.1 | 8 | 1533 | |||
XM_005265671.3 | XP_005265728.1 | 8 | 1877 | |||
XM_005265672.3 | XP_005265729.1 | 8 | 1877 | |||
XM_005265673.3 | XP_005265730.1 | 8 | 1877 | |||
XM_017008011.1 | XP_016863500.1 | 8 | 1108 | |||
XM_017008012.1 | XP_016863501.1 | 8 | 1108 | |||
XM_017008013.1 | XP_016863502.1 | 8 | 1108 | |||
XM_017008014.1 | XP_016863503.1 | 8 | 1877 | |||
XM_017008015.1 | XP_016863504.1 | 8 | 1108 | |||
XM_017008016.1 | XP_016863505.1 | 8 | 1877 | |||
XM_017008017.1 | XP_016863506.1 | 8 | 1108 | |||
GenBank mRNA | AB014597.2 | - | 8 | 1057 | ||
AF308285.1 | - | 8 | 1520 | |||
AK299046.1 | - | 3 | 225 | |||
AK300302.1 | - | 8 | 1107 | |||
AK307581.1 | - | 8 | 1533 | |||
AL833287.1 | - | 5 | 710 | |||
BC004173.2 | - | 8 | 1514 | |||
BC007747.2 | - | 6 | 863 | |||
BC043394.1 | - | 8 | 1502 | |||
BX648035.1 | - | 8 | 1106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
