siRNA Details
Gene Symbol: RGS22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286692.1 | NP_001273621.1 | 21 | 3213 | ||
NM_001286693.1 | NP_001273622.1 | 19 | 3108 | |||
NM_015668.4 | NP_056483.3 | 21 | 3249 | |||
XM_005250856.2 | XP_005250913.1 | 20 | 2936 | |||
XM_005250857.2 | XP_005250914.1 | 21 | 3095 | |||
XM_006716540.2 | XP_006716603.1 | 21 | 3363 | |||
XM_011516957.1 | XP_011515259.1 | 22 | 3544 | |||
XM_011516958.1 | XP_011515260.1 | 21 | 3442 | |||
XM_017013309.1 | XP_016868798.1 | 20 | 2936 | |||
XM_017013311.1 | XP_016868800.1 | 14 | 1792 | |||
GenBank mRNA | AK097399.1 | - | 10 | 1282 | ||
AK292559.1 | - | 21 | 3059 | |||
AL117544.1 | - | 2 | 74 | |||
AY009106.1 | - | 21 | 3042 | |||
BC036665.1 | - | 21 | 3100 | 5268331 | ||
BC092411.1 | - | 21 | 3246 | 30386231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
