siRNA Details
Gene Symbol: RGS22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286692.1 | NP_001273621.1 | 24 | 3697 | ||
NM_001286693.1 | NP_001273622.1 | 22 | 3592 | |||
NM_015668.4 | NP_056483.3 | 24 | 3733 | |||
XM_005250856.2 | XP_005250913.1 | 23 | 3420 | |||
XM_005250857.2 | XP_005250914.1 | 23 | 3420 | |||
XM_005250858.2 | XP_005250915.1 | 23 | 3417 | |||
XM_006716540.2 | XP_006716603.1 | 24 | 3847 | |||
XM_011516957.1 | XP_011515259.1 | 25 | 4028 | |||
XM_011516958.1 | XP_011515260.1 | 24 | 3926 | |||
XM_017013309.1 | XP_016868798.1 | 22 | 3261 | |||
XM_017013310.1 | XP_016868799.1 | 22 | 3258 | |||
XM_017013311.1 | XP_016868800.1 | 17 | 2276 | |||
GenBank mRNA | AK097399.1 | - | 13 | 1766 | ||
AK292559.1 | - | 24 | 3543 | |||
AL117544.1 | - | 5 | 558 | |||
BC036665.1 | - | 24 | 3584 | 5268331 | ||
BC092411.1 | - | 24 | 3730 | 30386231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
