siRNA Details
Gene Symbol: ATP13A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024524.3 | NP_078800.3 | 13 | 1831 | ||
XM_005269357.3 | XP_005269414.1 | 13 | 1852 | |||
XM_011513120.2 | XP_011511422.1 | 14 | 1953 | |||
XM_011513121.1 | XP_011511423.1 | 13 | 1831 | |||
XM_011513122.1 | XP_011511424.1 | 13 | 1525 | |||
XM_011513123.2 | XP_011511425.1 | 14 | 1958 | |||
XM_011513124.2 | XP_011511426.1 | 14 | 1958 | |||
GenBank mRNA | AJ306929.1 | - | 4 | 2355 | ||
AK024639.1 | - | 2 | 228 | |||
AK225064.1 | - | 2 | 228 | |||
DR005713.1 | - | 6 | 712 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
