siRNA Details
Gene Symbol: DOCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004946.2 | NP_004937.1 | 17 | 1665 | ||
XM_005265830.3 | XP_005265887.1 | 17 | 1665 | |||
XM_011534448.2 | XP_011532750.1 | 17 | 1665 | |||
XM_011534449.2 | XP_011532751.1 | 17 | 1665 | |||
XM_011534450.2 | XP_011532752.1 | 17 | 1665 | |||
XM_011534451.2 | XP_011532753.1 | 17 | 1665 | |||
XM_017009189.1 | XP_016864678.1 | 17 | 1665 | |||
XM_017009190.1 | XP_016864679.1 | 17 | 1665 | |||
GenBank mRNA | BC016996.1 | - | 4 | 302 | ||
BC104900.1 | - | 17 | 1621 | 8143903 | ||
BC113457.1 | - | 17 | 1621 | 8322509 | ||
BC143358.1 | - | 17 | 1621 | |||
D86964.1 | - | 17 | 1623 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
