siRNA Details
Gene Symbol: DDX60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017631.5 | NP_060101.3 | 34 | 4857 | ||
XM_011532103.2 | XP_011530405.1 | 34 | 4857 | |||
GenBank mRNA | AK000042.1 | - | 6 | 658 | ||
AK026333.1 | - | 2 | 123 | |||
AK223184.1 | - | 6 | 658 | |||
AK225191.1 | - | 6 | 645 | |||
AL137464.1 | - | 1 | 43 | |||
BC020601.1 | - | 2 | 73 | |||
BC038115.2 | - | 34 | 4802 | 5261001 | ||
BC074781.2 | - | 6 | 636 | |||
BX648655.1 | - | 14 | 1699 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
