siRNA Details
Gene Symbol: HECTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015382.3 | NP_056197.3 | 23 | 4169 | ||
XM_005267502.2 | XP_005267559.2 | 23 | 4331 | |||
XM_011536619.2 | XP_011534921.1 | 23 | 3767 | |||
XM_011536621.2 | XP_011534923.1 | 23 | 3767 | |||
XM_011536622.1 | XP_011534924.1 | 14 | 2094 | |||
XM_017021148.1 | XP_016876637.1 | 22 | 3661 | |||
XM_017021149.1 | XP_016876638.1 | 22 | 3661 | |||
XM_017021150.1 | XP_016876639.1 | 22 | 3661 | |||
XM_017021151.1 | XP_016876640.1 | 23 | 4363 | |||
XM_017021152.1 | XP_016876641.1 | 23 | 4363 | |||
XM_017021153.1 | XP_016876642.1 | 23 | 3767 | |||
GenBank mRNA | AB032957.2 | - | 15 | 2315 | ||
AK095634.1 | - | 14 | 2062 | |||
AY254380.1 | - | 23 | 3969 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
