siRNA Details
Gene Symbol: HECTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015382.3 | NP_056197.3 | 25 | 4570 | ||
XM_005267502.2 | XP_005267559.2 | 25 | 4732 | |||
XM_011536619.2 | XP_011534921.1 | 25 | 4168 | |||
XM_011536622.1 | XP_011534924.1 | 16 | 2495 | |||
XM_017021150.1 | XP_016876639.1 | 24 | 4062 | |||
GenBank mRNA | AB032957.2 | - | 17 | 2716 | ||
AK024847.1 | - | 3 | 399 | |||
AY254380.1 | - | 25 | 4370 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_144788, AK173106, AK141970.
Gene Ontology Categories:
