siRNA Details
Gene Symbol: HECTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015382.3 | NP_056197.3 | 25 | 4753 | ||
XM_005267502.2 | XP_005267559.2 | 25 | 4915 | |||
XM_011536619.2 | XP_011534921.1 | 25 | 4351 | |||
XM_011536622.1 | XP_011534924.1 | 16 | 2678 | |||
XM_017021148.1 | XP_016876637.1 | 24 | 4104 | |||
XM_017021149.1 | XP_016876638.1 | 24 | 4104 | |||
XM_017021150.1 | XP_016876639.1 | 24 | 4245 | |||
GenBank mRNA | AB032957.2 | - | 17 | 2899 | ||
AK024847.1 | - | 3 | 582 | |||
AK095634.1 | - | 16 | 2505 | |||
AY254380.1 | - | 25 | 4553 | |||
AY739714.1 | - | 1 | 70 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
