siRNA Details
Gene Symbol: EPB41L4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022140.3 | NP_071423.3 | 15, 16 | 1655 | ||
XM_011543530.1 | XP_011541832.1 | 15, 16 | 1832 | |||
XM_011543531.2 | XP_011541833.1 | 15, 16 | 1832 | |||
XM_011543532.1 | XP_011541834.1 | 15, 16 | 1832 | |||
XM_011543533.1 | XP_011541835.1 | 14, 15 | 1748 | |||
XM_017009689.1 | XP_016865178.1 | 14, 15 | 1748 | |||
GenBank mRNA | AB030240.1 | - | 15, 16 | 1387 | ||
BC114632.1 | - | 15, 16 | 1481 | |||
BC114942.1 | - | 15, 16 | 1476 | |||
CB048933.1 | - | 2, 3 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
