siRNA Details
Gene Symbol: EPB41L4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022140.3 | NP_071423.3 | 16, 17 | 1703 | ||
XM_011543530.1 | XP_011541832.1 | 16, 17 | 1880 | |||
XM_011543531.2 | XP_011541833.1 | 16, 17 | 1880 | |||
XM_011543532.1 | XP_011541834.1 | 16, 17 | 1880 | |||
XM_011543533.1 | XP_011541835.1 | 15, 16 | 1796 | |||
XM_017009689.1 | XP_016865178.1 | 15, 16 | 1796 | |||
GenBank mRNA | AB030240.1 | - | 16, 17 | 1435 | ||
AK096057.1 | - | 5, 6 | 385 | |||
BC114632.1 | - | 16, 17 | 1529 | |||
BC114942.1 | - | 16, 17 | 1524 | |||
CB048933.1 | - | 3, 4 | 258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
