siRNA Details
Gene Symbol: LMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022773.2 | NP_073610.2 | 9, 10 | 1428 | ||
NR_033645.1 | 10, 11 | 1557 | ||||
NR_033646.1 | 11, 12 | 1735 | ||||
XM_006720928.2 | XP_006720991.1 | 9, 10 | 1701 | |||
XM_006720929.3 | XP_006720992.1 | 10, 11 | 1582 | |||
XM_011522617.2 | XP_011520919.1 | 7, 8 | 1077 | |||
XM_017023575.1 | XP_016879064.1 | 9, 10 | 1426 | |||
XM_017023576.1 | XP_016879065.1 | 8, 9 | 1192 | |||
XM_017023577.1 | XP_016879066.1 | 10, 11 | 1857 | |||
XM_017023578.1 | XP_016879067.1 | 8, 9 | 1277 | |||
XM_017023579.1 | XP_016879068.1 | 8, 9 | 1285 | |||
XM_017023580.1 | XP_016879069.1 | 11, 12 | 2007 | |||
GenBank mRNA | AB075873.1 | - | 9, 10 | 1412 | ||
AK022743.1 | - | 9, 10 | 1426 | |||
AK025955.1 | - | 9, 10 | 1360 | |||
AK093050.1 | - | 6, 7 | 935 | |||
AK125201.1 | - | 10, 11 | 1559 | |||
AK225065.1 | - | 9, 10 | 1426 | |||
AK225406.1 | - | 9, 10 | 1413 | |||
AL832177.1 | - | 3, 4 | 3046 | |||
BC010738.1 | - | 8, 9 | 1193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
