siRNA Details
Gene Symbol: LMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022773.2 | NP_073610.2 | 11 | 2553 | ||
NR_033645.1 | 12 | 2682 | ||||
NR_033646.1 | 13 | 2860 | ||||
XM_006720928.2 | XP_006720991.1 | 11 | 2826 | |||
XM_006720929.3 | XP_006720992.1 | 12 | 2707 | |||
XM_011522617.2 | XP_011520919.1 | 9 | 2202 | |||
XM_017023576.1 | XP_016879065.1 | 10 | 2317 | |||
XM_017023577.1 | XP_016879066.1 | 12 | 2982 | |||
XM_017023578.1 | XP_016879067.1 | 10 | 2402 | |||
XM_017023579.1 | XP_016879068.1 | 10 | 2410 | |||
XM_017023580.1 | XP_016879069.1 | 13 | 3132 | |||
GenBank mRNA | AK022743.1 | - | 11 | 2551 | ||
AK093050.1 | - | 8 | 2060 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
