siRNA Details
Gene Symbol: LMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022773.2 | NP_073610.2 | 1 | 78 | ||
NR_033646.1 | 1 | 78 | ||||
XM_006720929.3 | XP_006720992.1 | 1 | 77 | |||
XM_011522614.2 | XP_011520916.1 | 1 | 76 | |||
XM_011522615.1 | XP_011520917.1 | 1 | 76 | |||
XM_011522618.2 | XP_011520920.1 | 1 | 76 | |||
XM_017023575.1 | XP_016879064.1 | 1 | 76 | |||
XM_017023578.1 | XP_016879067.1 | 1 | 76 | |||
XM_017023581.1 | XP_016879070.1 | 1 | 76 | |||
GenBank mRNA | AB075873.1 | - | 1 | 62 | ||
AK022743.1 | - | 1 | 76 | |||
AK093050.1 | - | 1 | 55 | |||
AK225065.1 | - | 1 | 76 | |||
AK225406.1 | - | 1 | 63 | |||
AK294329.1 | - | 1 | 76 | |||
AK294932.1 | - | 1 | 57 | |||
DA200865.1 | - | 1 | 78 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
