siRNA Details
Gene Symbol: QRICH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032134.2 | NP_115510.1 | 4 | 3328 | ||
XM_005257728.3 | XP_005257785.1 | 4 | 4338 | |||
XM_006722136.3 | XP_006722199.1 | 1 | 527 | |||
XM_011525344.2 | XP_011523646.1 | 2 | 3011 | |||
XM_017025206.1 | XP_016880695.1 | 4 | 4338 | |||
XM_017025207.1 | XP_016880696.1 | 3 | 4278 | |||
GenBank mRNA | AK058102.1 | - | 1 | 172 | ||
AL136774.1 | - | 4 | 3328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
