siRNA Details
Gene Symbol: QRICH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032134.2 | NP_115510.1 | 6 | 3526 | ||
NR_130649.1 | 6 | 617 | ||||
XM_005257728.3 | XP_005257785.1 | 6 | 4536 | |||
XM_006722136.3 | XP_006722199.1 | 2 | 639 | |||
XM_011525344.2 | XP_011523646.1 | 4 | 3209 | |||
XM_017025206.1 | XP_016880695.1 | 6 | 4536 | |||
XM_017025207.1 | XP_016880696.1 | 5 | 4476 | |||
XM_017025208.1 | XP_016880697.1 | 4 | 607 | |||
GenBank mRNA | AK058102.1 | - | 3 | 370 | ||
AL136774.1 | - | 6 | 3526 | |||
BC131559.1 | - | 6 | 502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
