siRNA Details
Gene Symbol: SIPA1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020808.4 | NP_065859.3 | 21 | 5403 | ||
XM_005273211.2 | XP_005273268.1 | 23 | 5740 | |||
XM_005273212.4 | XP_005273269.1 | 23 | 5547 | |||
XM_005273213.4 | XP_005273270.1 | 22 | 5440 | |||
XM_011544243.2 | XP_011542545.1 | 24 | 5596 | |||
XM_017001896.1 | XP_016857385.1 | 23 | 5493 | |||
XM_017001897.1 | XP_016857386.1 | 22 | 5686 | |||
XM_017001898.1 | XP_016857387.1 | 22 | 5460 | |||
GenBank mRNA | AB037810.1 | - | 21 | 4607 | ||
AK022852.1 | - | 12 | 1949 | |||
AK026779.1 | - | 8 | 1376 | |||
AK074212.1 | - | 6 | 836 | |||
AK093191.1 | - | 14 | 2485 | |||
AY168879.1 | - | 21 | 5363 | |||
BC013119.1 | - | 12 | 1860 | |||
BC082251.1 | - | 12 | 1889 | |||
BC150334.1 | - | 21 | 4618 | 8860510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
