siRNA Details
Gene Symbol: MAVS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020746.4 | NP_065797.2 | 2, 3 | 284 | ||
NR_037921.1 | 2, 3 | 284 | ||||
GenBank mRNA | AB033097.1 | - | 1, 2 | 190 | ||
AB097003.1 | - | 2, 3 | 247 | |||
AB232371.1 | - | 1, 2 | 184 | |||
AK023799.1 | - | 2, 3 | 240 | |||
AK123956.1 | - | 2, 3 | 245 | |||
AK130992.1 | - | 2, 3 | 205 | |||
AK296897.1 | - | 2, 3 | 249 | |||
BC044952.1 | - | 2, 3 | 218 | 5751684 | ||
BM834250.1 | - | 2, 3 | 248 | |||
DB219666.1 | - | 2, 3 | 284 | |||
DQ167126.1 | - | 1, 2 | 112 | |||
DQ174270.1 | - | 1, 2 | 112 | |||
DQ181928.1 | - | 1, 2 | 112 | |||
EF467324.1 | - | 1, 2 | 112 | |||
KC415005.1 | - | 1, 2 | 112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
