siRNA Details
Gene Symbol: SCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320044.1 | NP_001306973.1 | 19 | 3265 | ||
NM_012235.3 | NP_036367.2 | 22 | 3753 | |||
XM_005264967.1 | XP_005265024.1 | 22 | 3677 | |||
XM_005264968.1 | XP_005265025.1 | 19 | 3422 | |||
XM_005264971.1 | XP_005265028.1 | 16 | 2833 | |||
XM_011533501.1 | XP_011531803.1 | 22 | 3780 | |||
XM_011533502.2 | XP_011531804.1 | 14 | 2429 | |||
XM_017005918.1 | XP_016861407.1 | 22 | 3767 | |||
XM_017005919.1 | XP_016861408.1 | 22 | 3700 | |||
XM_017005920.1 | XP_016861409.1 | 22 | 3697 | |||
XM_017005921.1 | XP_016861410.1 | 16 | 2830 | |||
GenBank mRNA | AK027402.1 | - | 16 | 2743 | ||
AK075528.1 | - | 10 | 923 | |||
AK091005.1 | - | 1 | 1585 | |||
AK291484.1 | - | 17 | 2943 | |||
AK300311.1 | - | 19 | 3155 | |||
BC020987.1 | - | 14 | 2471 | 3852258 | ||
BC027207.2 | - | 17 | 2446 | |||
D83782.2 | - | 22 | 3739 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
