siRNA Details
Gene Symbol: SCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320044.1 | NP_001306973.1 | 17, 18 | 3084 | ||
NM_012235.3 | NP_036367.2 | 20, 21 | 3572 | |||
XM_005264967.1 | XP_005265024.1 | 20, 21 | 3496 | |||
XM_005264968.1 | XP_005265025.1 | 17, 18 | 3241 | |||
XM_005264971.1 | XP_005265028.1 | 14, 15 | 2652 | |||
XM_011533501.1 | XP_011531803.1 | 20, 21 | 3599 | |||
XM_011533502.2 | XP_011531804.1 | 12, 13 | 2248 | |||
XM_017005918.1 | XP_016861407.1 | 20, 21 | 3586 | |||
XM_017005919.1 | XP_016861408.1 | 20, 21 | 3519 | |||
XM_017005920.1 | XP_016861409.1 | 20, 21 | 3516 | |||
XM_017005921.1 | XP_016861410.1 | 14, 15 | 2649 | |||
GenBank mRNA | AK027402.1 | - | 14, 15 | 2562 | ||
AK075528.1 | - | 8, 9 | 742 | |||
AK291484.1 | - | 15, 16 | 2762 | |||
AK300311.1 | - | 17, 18 | 2974 | |||
BC020987.1 | - | 12, 13 | 2290 | 3852258 | ||
BC027207.2 | - | 15, 16 | 2265 | |||
D83782.2 | - | 20, 21 | 3558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
