siRNA Details
Gene Symbol: SCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320044.1 | NP_001306973.1 | 20 | 3392 | ||
NM_012235.3 | NP_036367.2 | 23 | 3880 | |||
XM_005264967.1 | XP_005265024.1 | 23 | 3804 | |||
XM_005264968.1 | XP_005265025.1 | 20 | 3549 | |||
XM_005264971.1 | XP_005265028.1 | 17 | 2960 | |||
XM_011533501.1 | XP_011531803.1 | 23 | 3907 | |||
XM_011533502.2 | XP_011531804.1 | 15 | 2556 | |||
XM_017005918.1 | XP_016861407.1 | 23 | 3894 | |||
XM_017005919.1 | XP_016861408.1 | 23 | 3827 | |||
XM_017005920.1 | XP_016861409.1 | 23 | 3824 | |||
XM_017005921.1 | XP_016861410.1 | 17 | 2957 | |||
GenBank mRNA | AK027402.1 | - | 17 | 2870 | ||
AK075528.1 | - | 11 | 1050 | |||
AK091005.1 | - | 1 | 1792 | |||
AK291484.1 | - | 18 | 3070 | |||
AK300311.1 | - | 20 | 3282 | |||
BC020987.1 | - | 15 | 2598 | 3852258 | ||
BC027207.2 | - | 18 | 2573 | |||
BG179218.1 | - | 2 | 125 | |||
D83782.2 | - | 23 | 3866 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
