siRNA Details
Gene Symbol: NME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018137.2 | NP_001018147.1 | 4, 5 | 600 | ||
NM_001018138.1 | NP_001018148.1 | 3, 4 | 458 | |||
NM_001018139.2 | NP_001018149.1 | 4, 5 | 416 | |||
NM_002512.3 | NP_002503.1 | 4, 5 | 579 | |||
GenBank mRNA | BC002476.2 | - | 4, 5 | 381 | 3349768 | |
BC095458.1 | - | 4, 5 | 390 | 30524221 | ||
BM556410.1 | - | 4, 5 | 416 | |||
BT007045.1 | - | 3, 4 | 349 | |||
CR541718.1 | - | 3, 4 | 349 | |||
EU794639.1 | - | 4, 5 | 398 | |||
L16785.1 | - | 4, 5 | 398 | |||
M36981.1 | - | 4, 5 | 421 | |||
X58965.1 | - | 4, 5 | 421 |
Gene Symbol: NME1-NME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018136.2 | NP_001018146.1 | 7, 8 | 825 | ||
NR_037149.1 | 8, 9 | 1045 | ||||
GenBank mRNA | AK313448.1 | - | 7, 8 | 782 | ||
BC107894.1 | - | 8, 9 | 947 | 6717551 | ||
BC133029.1 | - | 7, 8 | 728 | 40147051 | ||
BC133031.1 | - | 7, 8 | 728 | 40147053 | ||
DQ109675.1 | - | 7, 8 | 745 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008704, NM_008705, NM_001077529, AK216511, M65037, and others.
Gene Ontology Categories:
