siRNA Details
Gene Symbol: TTC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145418.1 | NP_001138890.1 | 7 | 2829 | ||
XM_005261405.2 | XP_005261462.1 | 7 | 2829 | |||
XM_006724171.3 | XP_006724234.1 | 6 | 11607 | |||
XM_011530018.2 | XP_011528320.1 | 7 | 3008 | |||
XM_011530019.2 | XP_011528321.1 | 7 | 2829 | |||
XM_011530020.1 | XP_011528322.1 | 7 | 2829 | |||
XM_011530021.2 | XP_011528323.1 | 7 | 2829 | |||
XM_011530022.1 | XP_011528324.1 | 7 | 2829 | |||
XM_017028673.1 | XP_016884162.1 | 7 | 2802 | |||
GenBank mRNA | AB028966.3 | - | 2 | 1622 | ||
AB665284.1 | - | 7 | 2777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
