siRNA Details
Gene Symbol: FRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023037.2 | NP_075463.2 | 14 | 1923 | ||
XM_006719749.3 | XP_006719812.1 | 14 | 3004 | |||
XM_017020303.1 | XP_016875792.1 | 14 | 3481 | |||
XM_017020304.1 | XP_016875793.1 | 14 | 1923 | |||
XM_017020305.1 | XP_016875794.1 | 14 | 3500 | |||
GenBank mRNA | AL049784.1 | - | 14 | 1923 | ||
U50534.1 | - | 14 | 1922 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
