siRNA Details
Gene Symbol: SGSM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098509.1 | NP_001091979.1 | 16 | 2124 | ||
NM_014853.2 | NP_055668.2 | 17 | 2261 | |||
XM_011524101.2 | XP_011522403.1 | 18 | 2337 | |||
XM_011524102.2 | XP_011522404.1 | 18 | 2335 | |||
XM_011524103.2 | XP_011522405.1 | 18 | 2336 | |||
XM_011524104.2 | XP_011522406.1 | 18 | 2861 | |||
XM_011524105.2 | XP_011522407.1 | 17 | 2201 | |||
XM_011524106.1 | XP_011522408.1 | 17 | 2159 | |||
XM_011524108.1 | XP_011522410.1 | 16 | 2024 | |||
XM_017025474.1 | XP_016880963.1 | 17 | 2199 | |||
XM_017025475.1 | XP_016880964.1 | 17 | 2200 | |||
XM_017025476.1 | XP_016880965.1 | 12 | 1496 | |||
GenBank mRNA | AB007857.3 | - | 16 | 2124 | ||
AB275762.1 | - | 17 | 2096 | |||
AB449878.1 | - | 16 | 1949 | |||
AK124883.1 | - | 4 | 2802 | |||
AK125203.1 | - | 4 | 1866 | |||
AK294955.1 | - | 16 | 2016 | |||
BC039204.1 | - | 17 | 2261 | 4817100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
